Neon Wand
# de catalogue 470201-082
Fournisseur: GSC International
Spécifications
- Description:Neon Wand
- Supplier No.:1401
Spécifications
A propos de cet article
Glows with Characteristic Brilliance!
- Contains neon at reduced pressure.
The neon gas glows when the tube is held in, or passed through a high frequency field, or a high voltage field, even though the tube has no electrodes.
Please Note: Not recommended for use with anything that produces a voltage higher than 75,000 volts.
Customers Who Bought This Also Bought