Ward's® Prepared Nematode Growth Medium
# de catalogue 470180-736
Fournisseur: IPM SCIENTIFIC, INC.
Spécifications
- Description:Nematode growth medium
- Packaging:Bottle
- Supplier No.:WACP470180-736
Spécifications
A propos de cet article
Designed for cultivation of Ceanorhabditis elegans.
- Sterile - no autoclaving
- Convenient - prepared for you
- Prepared consistenly every time
Prepared nematode growth medium, available as sterile agar plates and agar bottles. Great for aseptic culturing of Caenorhabditis elegans.
Ordering information: During checkout, please specify a delivery date for your specimens to arrive.
Delivery: Prepared plates will ship via overnight or 2nd day delivery, prepared bottles do not require as much temperature regulation in-transit so we ship them via ground to save you the cost of shipping.
Customers Who Bought This Also Bought