Natural Crystal Collection
# de catalogue 470112-330
Fournisseur: SCOTT RESOURCES 2, LLC SE
Spécifications
- Description:Natural crystal collection
- Cat. no.:470112-330
- Supplier No.:2305
Spécifications
A propos de cet article
Identify all six crystal systems (cubic, tetragonal, orthorhombic, monoclinic, triclinic and hexagonal) with the 15 representative specimens of natural crystals included in this collection. In compartmented box with key sheet listing the locality and crystal form of each specimen.
Customers Who Bought This Also Bought