Muskrat Skull, Ward’s®
# de catalogue 470027-956
Fournisseur: Avantor
Spécifications
- Description:Muskrat Skull
- Height:1.5"
- Cat. no.:470027-956
- Length:2 .5"
- Width:1.5"
- Supplier No.:WARD470027-956
- No. of Parts:1
Spécifications
A propos de cet article
Study the greatly enlarged incisors of Ondatra zibethicus with the unmounted skull specimen.
- Natural bone
- Professionally cleaned and prepared
Ordering information: This is a real specimen, so size and appearance may vary from the image.
Customers Who Bought This Also Bought