Muscle Action Demonstration Model
# de catalogue 470019-326
Fournisseur: Avantor
Spécifications
- Description:Muscle Action Demonstration
- Cat. no.:470019-326
- Supplier No.:WARD470019-326
Spécifications
A propos de cet article
Unique Apparatus Reproduces The Movements Of Students’ Own Muscles
By using this interactive model, students better understand the physics behind how muscles work to create movement. Velcro straps secure the apparatus to the student’s upper and lower arm. By straightening and bending their arm, the model mimics the opposing contractions and relaxations of their own biceps and triceps. The “bones” are constructed of durable plastic, while the “muscles” are composed of a tough, elastic material. It comes with instructions. Size: measures 8" from upper arm to elbow, 8 1/2 " from elbow to fingertips.
Customers Who Bought This Also Bought