Multipurpose Drawing Compass
# de catalogue 470122-024
Fournisseur: Educational Tools
Spécifications
- Description:Multipurpose drawing compass
- Cat. no.:470122-024
- Supplier No.:B-72-2
Spécifications
A propos de cet article
Consists of three 2" Ø, joined aluminum disks with knob tightening adjustment which holds pencils, chalk or hobby knives up to 0.6" in Ø. The diameter of the desired circle is set using scale on the front disk.
Ordering information: Supplied with pencil.
Customers Who Bought This Also Bought