ContactUsLinkComponent
Mueller-Hinton Agar
Mueller-Hinton Agar
# de catalogue 470180-638
Fournisseur:  IPM SCIENTIFIC, INC.
Numéro CAS:  
Mueller-Hinton Agar
# de catalogue 470180-638
Fournisseur:  IPM SCIENTIFIC, INC.
Numéro CAS:  
Spécifications
  • Description:
    Dehydrated Mueller-Hinton Agar
  • Cat. no.:
    470180-638
  • Supplier No.:
    880017
Spécifications
A propos de cet article
For testing antimicrobial susceptibility of microorganisms by agar diffusion method.
Premeasured Packs Save Time in the Lab
Simply add the contents of one Dehydrated Media pack to one liter of water, bring to a boil, and sterilize, to make enough media for 40 to 50 plates or 16edited0 tubes.
Customers Who Bought This Also Bought