A propos de cet article
The Moment of Force Apparatus contains everything needed to study principles of mass, levers, moments, forces, and equivalence.
This set contains everything needed to study important principles of mass, levers, moments, forces, and equivalence.
Determine the distance between the application of a force and the axis of rotation, and demonstrate that the moment of any force is the equal to the force times its lever arm.
The set includes a meter stick, three knife edge clamps, a heavy cast iron support, two 500 g capacity mass hangers and a set of eight slotted masses (10 g to 500 g) with a storage rack. Complete with instructions and suggestions for use.
- Three Knife-Edge Clamps
- Heavy, Cast-Iron Support
- Two 500 g Capacity Mass Hangers
- A Set of Eight Slotted Masses (10 g to 500 g) with a Storage Rack
- Instructions and Suggestions for Use Included
Users determine the distance between the application of a force and the axis of rotation.
The machine features three-knife edge clamps, a cast-iron support, two 500 g capacity mass hangers, a meter stick, and more.