Molecular Orbital Models
# de catalogue 470017-652
Fournisseur: United Scientific Supplies
Spécifications
- Description:Set of 7 Molecular Orbital Models
- Cat. no.:470017-652
- Supplier No.:MOMS07
Spécifications
A propos de cet article
The most common atomic and hybridized molecular orbitals are illustrated in this set of seven models.
- Models Mounted on Z-axis
- Removable
- Seven Models per Set
Each model, which is mounted on a removable stand, is assembled on a vertical rod that represents the Z-axis.
Models include the following orbitals: S, PX, PY, PZ, SP1, SP2, and SP3.
Customers Who Bought This Also Bought