Molecular Motion Tube
# de catalogue 470110-946
Fournisseur: Montclare
Spécifications
- Description:Molecular Motion Tube
- Cat. no.:470110-946
- Supplier No.:6003/PRINT
Spécifications
A propos de cet article
A smashing demonstration for the kinetic theory of matter. The 12" glass tube contains numerous colored glass particles and a small amount of mercury.
Because the tube is sealed off at reduced pressure, the mercury will boil when placed over a moderate heat source. As the tube is heated, the invisible mercury molecules violently agitate the visible glass particles, creating an effective display of random molecular motion.
Caution: Warning: This product contains mercury and may not be available for sale in your state. Contact customer service for details.
Customers Who Bought This Also Bought