The Mole Box
# de catalogue 470012-918
Fournisseur: Science First
Spécifications
- Description:The Mole Box
- Cat. no.:470012-918
- Supplier No.:30-165
Spécifications
A propos de cet article
Visualize Volume of Moles
Imagine how much easier it would be if atoms were the size of basketballs and you could really show students what it was you were talking about when you lectured on Avogadro’s Number, relative molecular mass, molar volume and Gas Law. Well, this product is as close as you can get to helping students see atoms as large, tangible objects. The Mole Box is a 22.4 L cardboard cube that represents the equivilent of 1 mole of O2, N2, H2, and He at STP of 0°C, and 1 atm. It features Avogadro’s Number and the gas law with large, easy-to-read equations.
Customers Who Bought This Also Bought