Molarity Lab Activity
# de catalogue 470023-900
Fournisseur: Kemtec
Spécifications
- Description:Molarity Lab Activity
- No. of Lab Groups:4
- Cat. no.:470023-900
- Includes:Teacher's Guide and Student Copymasters
- Supplier No.:13-136
Spécifications
A propos de cet article
Use the Mole Concept to Calculate Various Concentrations of Chemical Solutions.
- Only requires basic labware
- Determine unknown concentrations using titrations
Determine the concentrations of four solutions of hydrochloric acid in unknown concentrations by titrating them with known concentrations of sodium hydroxide. The kit contains two laboratory exercises, a reproducible manual and enough materials for four setups.
Customers Who Bought This Also Bought