Oven Bake Modeling Clay
# de catalogue 470102-606
Fournisseur: POLYFORM PRODUCTS COMPANY
Spécifications
- Color:White, purple, green, orange, black, brown, yellow, red, blue, and beige
- Description:Modeling Clay, Basic Colors
- Cat. no.:470102-606
- Supplier No.:S3MP00001
Spécifications
A propos de cet article
Prepare designs easily without premature hardening.
- Non-toxic
- Soft and pliable
Can be tooled, rolled, cut, extruded, or formed over molds. To harden, bake in a conventional oven at 275° for 15 minutes. Once cooled, the baked pieces can be polished, drilled, sanded, carved, and painted.
Delivery: Set contains (10) 2 oz./57 g bars of 10 different colors.
Caution: Conventional oven only. Do not microwave.
Customers Who Bought This Also Bought