Mobile Metal Cabinet
# de catalogue 470022-506
Fournisseur: LYON, LLC.
Spécifications
- Description:Mobile metal cabinet
- Cat. no.:470022-506
- Supplier No.:MOBILECAB4DRWR
Spécifications
A propos de cet article
In addition to its mobility, with its 5" swivel casters, this mobile cabinet's parts are versatile as well.
- Removable steel tray for even more storage and totability
- Individual drawer locks give you access to some materials, while providing security for others
The reversible top acts as a countertop or tray with ¹/₂" flange; inside each drawer is a sliding. The steel cabinet is finished in gray, with four deep drawers, 21³/₄×20"×6³/₄" Ext.Ø. each. Powder paint finish. Assembly is required; it comes with instructions. Overall size: 22¹/₄W x 20¹/₂D x 34"H.
Weight: 135 lbs.
Customers Who Bought This Also Bought