Mobile Demonstration Table
# de catalogue 470002-758
Fournisseur: Diversified Woodcrafts
Spécifications
- Base/Leg Material:Red oak
- Worksurface Material:ChemGuard®
- Description:Mobile demonstration table with built-in sink
- Height:36"
- Cat. no.:470002-758
- W×D:48×28"
- Weight:215 lbs.
- Supplier No.:4222K
Spécifications
A propos de cet article
A self contained demonstration center with its own water supply, waste container, stainless steel sink and hand pump.
- Constructed of solid oak and oak hardwood veneers
- Finished in a non-emitting, chemical resistant UV finish
The four drawers are mounted on epoxy-coated nylon bearing glides. Unbreakable aluminium sliding/locking doors conceal the drawers and storage compartment, and the supplied heavy-duty 4" ball-bearing swivel casters with brakes support 500 lbs. The top is 1¼" thick ChemGuard high pressure laminate.
Sink WxLxD: 15×13×5"
Certifications: SEFA compliant and MAS Certified Green.
Ordering information: Unit is also available without the sink, faucet, rod sockets and upright set.
Delivery: Includes a GFI protected AC duplex electrical outlet and can be powered with the supplied 25' extension cord, two 2½ gallon water bottles, two ¾" diameter upright rods, one ½" diameter crossbar and clamps.
Customers Who Bought This Also Bought