Ward's® Mitotic Stage Counts Flashcards
# de catalogue 470022-904
Fournisseur: PROFORMA ASSOCIATES
Spécifications
- Description:Mitotic Counts Flash Cards, 25 Cards
- Cat. no.:470022-904
- Length:12.7 cm
- Width:7.6 cm
- Supplier No.:323378
Spécifications
A propos de cet article
Wide Field of View Shows Many Cells Captured at Various Phases of Mitosis
- Flashcards are the perfect supplement to any lab that requires the observation of plant mitosis
Use the color photos of plant root tip mitosis to identify and quantify the different stage of mitosis shown on each card and to determine the relative duration for each stage.
Customers Who Bought This Also Bought