ContactUsLinkComponent
Mini-Prep Isolation of Plasmid DNA
Mini-Prep Isolation of Plasmid DNA
# de catalogue 470121-614
Fournisseur:  Edvotek
MINI-PREP ISOLATION OF PLASMID
Mini-Prep Isolation of Plasmid DNA
# de catalogue 470121-614
Fournisseur:  Edvotek
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Mini-Prep Isolation of Plasmid DNA
  • Cat. no.:
    470121-614
  • Supplier No.:
    202-B
Spécifications
A propos de cet article
Allow students to practice small-scale rapid isolation of plasmid DNA, which is a routine procedure used for screening and analysis of recombinant DNA in cloning and subcloning experiments. In this experiment, students isolate plasmid DNA in just 60 minutes, without the use of toxic chemicals such as phenol or chloroform.
Material provided can support up to 40 isolations and five DNA agarose gels. Kit includes instructions, Plasmid Extraction LyphoCells, gel loading solution, UltraSpec-Agarose powder, electrophoresis buffer, InstaStain FlashBlue and FlashBlue Plus stain, calibrated pipet, 100 ml graduated cylinder and microtipped transfer pipettes. Time requirements: 60 min isolation of plasmid; 45 min electrophoresis.
Ordering information: An electrophoresis tank, power supply, variable micropipet and tips, balance and microwave or hot plate are required and available seperately.
Customers Who Bought This Also Bought