Spécifications
- Description:Mini Hoffman Electrolysis Apparatus
- Cat. no.:470005-464
- Supplier No.:EQ116
Spécifications
A propos de cet article
Very Effective, Economical Alternative
A low cost electrolysis kit, this apparatus is designed to replace any cumbersomely large, fragile, and expensive models. Using this small-scale model, you can perform many different experiments simultaneously. Includes an electrolysis cell, pair of plastic stop cocks, pair of platinum electrodes, one roll transparent metric tape, and suggested experiment outlines.