ContactUsLinkComponent
Mini Demo Electric Motor
Mini Demo Electric Motor
# de catalogue 470014-344
Fournisseur:  VWR International
MOTOR DEMO ELECTRIC MINI
Mini Demo Electric Motor
# de catalogue 470014-344
Fournisseur:  VWR International
Spécifications
  • Description:
    Mini Demo Electric Motor
  • Cat. no.:
    470014-344
  • Supplier No.:
    PH1238
Spécifications
A propos de cet article
Economical and effective.
  • Simplest DC electric motor model available to education
This unit features a two pole armature wound with enameled copper wire. A permanent magnetic field is produced by a removable bar magnet. A disc type commutator is incorporated and external connection to the phosphor-bronze brushes is through of a pair of 4 mm sockets. Unit operates on 6–8 volts DC. required but not included is a power supply and connecting wires.
Customers Who Bought This Also Bought