ContactUsLinkComponent
Mini Buzzer
Mini Buzzer
# de catalogue 470102-652
Fournisseur:  Ajax Scientific
BUZZER MINI 12 V W/ LEADS 7-16 V
Mini Buzzer
# de catalogue 470102-652
Fournisseur:  Ajax Scientific
Spécifications
  • Description:
    Buzzer, Mini, 3-24V with Leads
  • Cat. no.:
    470102-652
  • Supplier No.:
    16-138
Spécifications
A propos de cet article
Mini Buzzer with 2 leads. 3 to 24 V.
Customers Who Bought This Also Bought