Mineral ID Lab with Sort Guide
# de catalogue 470151-136
Fournisseur: Geoscience Industries
Spécifications
- Description:Mineral ID Lab With Sort Guide
- No. of Samples:20
- Cat. no.:470151-136
- Supplier No.:1761-100
Spécifications
A propos de cet article
Teach your students how to detect various minerals by their hardness, luster, streak and color.
Contains 20 minerals, streak plate, iron nail, magnifier and magnet. Also includes Sort Guides to help you accurately identify the sample.
Customers Who Bought This Also Bought