Spécifications
- Description:Mimio teach interactive system
- Cat. no.:470017-940
- Supplier No.:1762261
Spécifications
A propos de cet article
Add Interactive to Your Current Dry Erase Board
Transform any dry erase board into a fully interactive whiteboard with this compact classroom technology solution. The patented infrared and ultrasound sensor technology fits neatly into a small, unobtrusive bar that attaches easily to a board. Combine it with a projector and computer, and it becomes a fully interactive system with all the tools of an interactive whiteboard in a budget-friendly package. Lightweight yet durable, the MimioTeach system is easy to handle and transport, plus it mounts to your current whiteboard magnetically - no need for tricky installation. Powerful software drives the system and dramatically expands teaching options to encourage active learning. Interactive mode with projector supports whiteboard sizes of 6.6 ft x 7.9 ft., up to 10 ft. in standard aspect ratio diagonal projection display or whiteboards up to 9.4 ft. diagonal in wide screen mode. This comprehensive package includes MimioTeach bar with embedded wireless interface, rechargeable stylus, MimioHub™ wireless receiver, USB micro-B cable (5 m/16 ft), magnetic mounting bracket, 5V 500 AUSB power supply, and a DVD containing Windows (MimioStudio 7) in 26 languages. The system comes with a 2 year warranty extended to 5 years on all hardware components except batteries, with online registration. Unlimited lifetime warranty on software. Battery warranty is one year from the date of shipment.