Edvotek® Micropipetting Basics
# de catalogue 470106-954
Fournisseur: Edvotek
Spécifications
- Description:Micropipetting Basics Educational Kit
- Cat. no.:470106-954
- Supplier No.:S-44
Spécifications
A propos de cet article
Teach your students how to use a micropipette with ease and accuracy with multicolored dyes. A fun and cost-effective way to learn this important skill.
- Complete in one class period (45 minutes)
- Includes instructions, various colored dye samples, and a Pipette Card™
- All you need is micropipette and tips
Store at room temperature.
Customers Who Bought This Also Bought