Ward's® Microfossil Strew Slide Set
# de catalogue 470026-772
Fournisseur: Avantor
Spécifications
- Description:Microfossil strew slide set
- Supplier No.:WARD470026-772
- No. of Parts:12 samples
Spécifications
A propos de cet article
View minute, fossilized life forms
- Made by Ward's geologists
- One of a kind paleontology slides
- Grade for elementary aged to college level students
What may appear to students as grains of sand are really the fossilized remains of marine organisms, including conodonts, forams, molluscs, and more. Samples, from various localities, span several geologic periods. A slide reference list is included. A hand lens or stereomicroscope, available separately, is recommended for best viewing.
Ordering information: Slide set includes 12 numbered cardboard microplanetary slides (1×3").
Customers Who Bought This Also Bought