Microbial Ecology in a Closed System Kit | Avantor
Microbial Ecology in a Closed System Kit
Microbial Ecology in a Closed System Kit, 30
Catalog # 470100-972
Microbial Ecology in a Closed System Kit
Catalog # 470100-972
Supplier:  
Microbial Ecology in a Closed System Kit
Microbial Ecology in a Closed System Kit
Microbial Ecology in a Closed System Kit, 30
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Design a Winogradsky Column
  • Demonstrate the use of a Winogradsky column
  • Study population succession
  • Includes a teacher's guide
Study population succession in an ecosystem filled with mud, shredded paper, salts and water. Use this kit to introduce students to the Winogradsky column, a small vertical pond designed to simulate various environmental conditions.
Includes materials for 30 students and a teacher’s guide with background information.
Spécifications
  • Désignation:
    Microbial Ecology in a Closed System Kit
  • No. of Lab Groups:
    30