Spécifications
- Description:Silicone Culture Gum
- Cat. no.:470024-704
- Grade:Laboratory
- Supplier No.:SS0133-30G
Spécifications
A propos de cet article
Construct an Aquarium on a Glass Microscope Slide
View “the world of the little” for an extended period of time without specimens drying up! WARD’S Culture Gum allows oxygen and waste gases to diffuse without evaporation. Simply use Culture Gum to construct the walls, add your microlife sample, and seal it with a coverslip. Our material is nontoxic, inert, reusable, and autoclavable. The kit comes with instructions, a list of activities, and one box each of 72 microscope slides and coverslips.