Metric/English Wind-Up Tapes
# de catalogue 470004-630
Fournisseur: American Scientific
Small Business Enterprise
Spécifications
- Length:1.5 m (5 ft.)
- Description:Wind-up tape, metric and imperial
- Cat. no.:470004-630
- Supplier No.:3967-00
Spécifications
A propos de cet article
These metric/English wind-up tapes, made of impact resistant plastic and featuring fiberglass tape, were designed to meet the rigors of the classroom.
Customers Who Bought This Also Bought