Metric Stacking Masses
# de catalogue 470110-708
Fournisseur: SI Manufacturing
Spécifications
- Description:Metric Stacking Masses
- Cat. no.:470110-708
- Supplier No.:91630
Spécifications
A propos de cet article
Designed to illustrate that height is proportional to mass, when diameter is uniform. Set consists of 52 identified masses which are molded to stack. Included are 4 yellow 10 g masses, eight red 5 g masses and 40 blue 1 g masses.
Customers Who Bought This Also Bought