Metric Micrometer
# de catalogue 470110-218
Fournisseur: Walter Stern
Spécifications
- Description:Metric Micrometer
- Cat. no.:470110-218
- Supplier No.:601A
Spécifications
A propos de cet article
Forged steel fork and hammertone finish make ideal for heavy use. Steel frame has satin chrome finished barrel with clearly graduated markings. Includes positive spindle lock and friction stop. Accuracy 0.076mm.
Includes positive spindle lock and friction stop.
Metric scale: 25mm.
Customers Who Bought This Also Bought