Meteorological Balloons
# de catalogue 470007-960
Fournisseur: Scientific Sales
Spécifications
- Description:Meteorological balloons, 6' dia.
- Cat. no.:470007-960
- Supplier No.:8233
Spécifications
A propos de cet article
Use the neoprene rubber balloons as a visual aid to help students determine cloud heights, wind speed, and wind direction.
- Each conforms to US Weather Bureau specifications
Customers Who Bought This Also Bought