Metal Storage Cabinets
Fournisseur: Tennsco
Certificats
A propos de cet article
Durable metal cabinets provide safe storage for labs and classrooms.
- Three-point locking mechanism
- Standard cabinet shelves hold up to 200 lbs.
- Reinforced doors for strength and quiet operation
- Adjustable shelving in 2" increments
These sturdy metal storage cabinets are available in five sizes to meet your lab and classroom needs. The cabinets feature two swing-out doors with 3-point locking mechanisms. Interior shelves can be adjusted in 2" increments to provide flexible storage. Storage capacity for Standard and Deluxe cabinets is 200 lbs. and capacity for Jumbo cabinets is 400 lbs.
The cabinet’s two adjustable front levelers keep it level on uneven floors. Standard Cabinet Shelves hold 200 lb., and Jumbo Cabinets hold up to 400 lb.
Ordering information: Please specify color choice when ordering.
Delivery: This ships fully assembled.
Customers Who Bought This Also Bought