Metal Madgeburg Hemispheres
# de catalogue 470039-952
Fournisseur: United Scientific Supplies
Spécifications
- Description:Metal Magdeburg Hemispheres
- Cat. no.:470039-952
- Supplier No.:MGH001
Spécifications
A propos de cet article
Hands on demomstration showing the power of a vacuum.
- Sturdy metal construction
- Easy to grab handles
Authentic metal construction makes these heavy duty hemispheres closer to the original version that plastic or rubber replicas. Their thick ircon walls and large steel handles deliver durability for years of student use. A brass plated valve and precision machining also prevent leakage.
Customers Who Bought This Also Bought