Metal Lab Storage Cabinet
# de catalogue 470022-490
Fournisseur: LYON, LLC.
Spécifications
- Description:Metal lab storage cabinet
- Cat. no.:470022-490
- Supplier No.:1081
Spécifications
A propos de cet article
The durable steel holds up to 180 lbs. per shelf, while nylon washers, plastic inserts, and felt pads create a quiet door closure system for the metal, lockable doors.
- Four shelves are easily adjustable to every 2"
- Dimension (W x D x H): 36×18×78"
- Weight: 129 lbs.
The putty-colored cabinet requires assembly and includes instructions.
Customers Who Bought This Also Bought