Marine Plankton Microslide
Fournisseur: SUPERTEK SCIENTIFIC LLC
Certificats
A propos de cet article
Investigate these Small Saltwater Organisms
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects it on both sides
Microslides consist of a strip of film with photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides. A detailed text folder is included. Viewers are available separately.
Ordering information: Set includes Diatoms(200X), Asterionella (200X), Gymnodinium (200X), Ceratium (30X), Foraminifera (20X), Radiolarians (30X), Zooplankton (50X), and Zoea larvae (25X).
Customers Who Bought This Also Bought