Map & Chart Cabinets
# de catalogue 470022-472
Fournisseur: FOSTER MANUFACTURING COMPANY
Spécifications
- Description:Base for small map and chart cabinet
- Cat. no.:470022-472
- Supplier No.:12808-GR
Spécifications
A propos de cet article
Extra-large drawers protect documents.
- Baked-on beige enamel finish inside and out
Provide optimum storage space for maps and charts, housing them flat to prevent damage. A precision ball bearing system allows you to easily slide drawers in and out, even when they’re filled to capacity, and safety stops keep the drawers from over extending. They are available in three different sizes, and you can use the five-drawer cabinets alone, stacked, or with a 7" base to raise them off the floor.
Customers Who Bought This Also Bought