Mammary Gland, Inactive, Human Slide
# de catalogue 470183-076
Fournisseur: Avantor
Spécifications
- Description:Mammary Gland, Inactive, Human
- Cat. no.:470183-076
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470183-076
- Orientation:Section
Spécifications
A propos de cet article
Female Reproductive Organ
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Abundant connective tissue is visible. The ducts and surrounding loose connective tissue constitute a lobule. The dense connective tissue contains aggregates of adipocytes.
Customers Who Bought This Also Bought