Magnetic Serial Number Restoration Kit
Fournisseur: PRECISION FORENSICS TESTING
Certificats
A propos de cet article
Study Forensics Applications for Magnetism
- Safe alternative to Chemical Alteration
- Reusable materials
Students will investigate the role of forensics in restoring serial numbers while learning about magnitism with this complete kit that includes all neccessary components . Without using any harsh chemicals, students can safely perform realistic techniques for serial number restoration right on their desktop, without the need for a fume hood. Materials supplied for four student groups.
Customers Who Bought This Also Bought