Magnetic Karyotyping Kit
# de catalogue 470015-312
Fournisseur: KELLER BROS. & MILLER, INC.
Spécifications
- Description:Magnetic Karyotyping Kit
- Cat. no.:470015-312
- Supplier No.:4627000
Spécifications
A propos de cet article
Large Photomicrographs of Chromosomes
Learn about human chromosomes quickly and easily with these convenient magnetic pieces. Simply arrange the chromosome spread on a magnetic board, then line up the homologous pairs in order of length and banding, creating a human karyotype large enough for the entire class to see. It can also be used to illustrate the genetic makeup of males and females, and genetic abnormalities including Down, Klinefelter, and Turner Syndromes. A human karyotype form, magnetic chromosomes, and instructions are all included.
Customers Who Bought This Also Bought