ContactUsLinkComponent
Magdeburg Hemispheres
Magdeburg Hemispheres
# de catalogue 470018-246
Fournisseur:  VWR International
MAGDEBURG HEMISPHERES PRESSURE DEMO
Magdeburg Hemispheres
# de catalogue 470018-246
Fournisseur:  VWR International
Spécifications
  • Description:
    Magdeburg Hemispheres
  • Cat. no.:
    470018-246
  • Supplier No.:
    PH0171A
Spécifications
A propos de cet article
Employs Guericke’s Method for Demonstrating the Force of Atmospheric Pressure.
  • Two plastic, 10cm-diameter hemispheres, each with a sturdy handle, make up this device
  • Operating instructions are included
One hemisphere has a gasket seal; the other has a stopcock for attaching to 0.8-1.2cm inner-diameter vacuum tubing. Join the hemispheres, hook them up to a hand pump, aspirator, or vacuum pump, and evacuate them. Students will find it hard, if not impossible, to pull them apart. Open the stopcock to let air in, and the hemispheres separate easily.
Customers Who Bought This Also Bought