Ward's® Prepared MacConkey Agar
# de catalogue 470180-742
Fournisseur: IPM SCIENTIFIC, INC.
Numéro CAS:
Spécifications
- Packaging:Plates
- Description:MacConkey Agar
- Cat. no.:470180-742
- Supplier No.:WARD470180-742
Spécifications
A propos de cet article
For Differential Isolation of Enteric Bacilli Based on Lactose Fermentation
- Sterile- no autoclaving
- Convenient - prepared for you
- Prepared consistenly every time
Ordering information: These products contain living or perishable materials
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought