The Lymphatic System Poster
# de catalogue 470112-284
Fournisseur: Anatomical Chart Co.
Spécifications
- Length:66 cm
- Width:50 cm
- Description:Lymphatic System Poster
- Cat. no.:470112-284
- Finish:Laminated
- Supplier No.:1587790254
Spécifications
A propos de cet article
Shows lymphatic system of stomach, pancreas, spleen and biliary tract.
Frontal view exposing superficial lymph glands, vessels, veins, and certain deep lymphatic structures.
Customers Who Bought This Also Bought