Prepared Luria Broth Media
Fournisseur: Avantor
Certificats
A propos de cet article
This high-nutrient broth is a great general purpose media. Especially useful for growing E. coli cultures
- Sterile, pre-prepared media; just inoculate and incubate
Each tube contains 9mL of liquid broth.
Each bottle contains 200mL of liquid broth.
Customers Who Bought This Also Bought