Ward's® Luria Agar Media Plates and Pour Packs
Supplier: IPM SCIENTIFIC, INC.
Certificates
About this item
All-purpose medium that supports the growth of a variety of microorganisms.
- Sterile, pre-prepared media; just inoculate and incubate
- All-purpose media
- Excellent for preparing streak plates to determine colony morphology
- Great for coliphage plaque assays
Choose between prepared media plates or pouring your own.
This nutrient rich broth is a non-selective media that is used to grow most types of aerobic organisms. Each bottle pours up to ten petri dishes. Simply melt in a microwave oven, water bath or autoclave. Once cool enough to handle, pour into sterile petri-dish plates.
Delivery: Small Pour Packs come with two, 200 ml bottles of the agar of your choice, and 20 standard (100 mm diameter) petri-dishes sterilized by gamma irradiation.
Large Pour Packs come with six, 200 ml bottles of the agar of your choice, and 60 standard (100 mm diameter) petri-dishes sterilized by gamma irradiation.
Large Pour Packs come with six, 200 ml bottles of the agar of your choice, and 60 standard (100 mm diameter) petri-dishes sterilized by gamma irradiation.
Customers Who Bought This Also Bought