Lung Volume Bag Sets
Fournisseur: Nasco
Certificats
A propos de cet article
This set is a simple and inexpensive tool that measures vital capacity.
- Set measures vital capacity
- Set also compares effects of height, age, smoking, and more
- Four mouthpieces included
- Simple
- Inexpensive
Vital capacity is the volume of air a person can expel after taking the deepest possible breath.
This set can also be used to compare effects of height, age, physical activity, and smoking.
Ordering information: This kit includes four holders, four mouthpieces, and four heavy-gauge plastic bags calibrated to 6 L.
Customers Who Bought This Also Bought