Low Frequency Generator
Catalog # 470018-260
Supplier: VWR International
Specifications
- Description:Low Frequency Generator
- Cat. no.:470018-260
- Supplier No.:PH1298-N9
Specifications
About this item
Reliable and User Friendly
- Frequency range : 1HZ-2 kHz.
- Waveform: sine square and triangular. TTL output: controllable ±13 Volts when auxiliary input of 15 Volts is used, 9 volts if batteries are used.
- Can use an external power supply or two 9V batteries, not included.
Two potentiometers for frequency and amplitude are featured on this generator along with two dials for selection of frequency and waveform. Two outputs - the alternating signal and the TTL signal are also included.
Customers Who Bought This Also Bought