Boyle's Law Apparatus
# de catalogue 470148-928
Fournisseur: VWR International
Spécifications
- Description:Boyle's Law Apparatus
- Cat. no.:470148-928
- Supplier No.:P12030
Spécifications
A propos de cet article
This apparatus makes it easy for students to learn about Boyle's Law.
- Calibrated Plastic Syringe
- Wooden Base
- Lubricated Piston
- Various Sizes of Weights
- Students Plot Pressure against the Volume of Gases in the Cylinder
The calibrated plastic syringe is anchored onto a wooden base.
A lubricated piston is mounted to a platform that holds various size of weights, so students can plot pressure against the volume of the gases in the cylinder.
Customers Who Bought This Also Bought