Live Restriction Enzymes
Supplier: Avantor
Certificates
About this item
These restriction enzymes are used to recognize specific sequences in double strand DNA. BamH I recognizes 5-G/GATCC-3, Ecor I recognizes 5-G/AATTC-3, and Hind III recognizes 5-A/AGCTT-3. The activity level of all our restriction enzymes is 10 units per 1 µl.
All are shipped next day on ice. As they are perishable, a ship date must be specified.
Customers Who Bought This Also Bought