Litmus Test Sheets
Fournisseur: Precision Laboratories
Certificats
A propos de cet article
These litmus test sheets are available in litmus red (ACS) or litmus blue (ACS).
- Individual Sheet Length: 8"
- Individual Width: 10"
- Twenty-Four Sheets in One Package
- Litmus Blue and Litmus Red Available
These individual sheets of litmus test papers are 8 x 10".
One package has 24 sheets.
Packaging: One package has 24 sheets.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.