Litmus Milk Broth
# de catalogue 470180-732
Fournisseur: Avantor
Numéro CAS:
Spécifications
- Description:Prepared Litmus Milk Broth Tubes
- Cat. no.:470180-732
- Supplier No.:WARD470180-732
Spécifications
A propos de cet article
For demonstration of acid production, coagulation, or proteolysis of casein in culture identification.
Customers Who Bought This Also Bought