Liter Cube
# de catalogue 470014-884
Fournisseur: SI Manufacturing
Spécifications
- Description:Liter Cube
- Cat. no.:470014-884
- Supplier No.:77864
Spécifications
A propos de cet article
Demonstrate the Relation of Length, Area, and Volume
- Graduated in 100 mL intervals
- Illustrate that volume = height x base
- Convenient for measuring mass
This clear, acrylic liter cube is graduated to 1,000 mL in 100 mL intervals and has a cm grid on the bottom to illustrate that volume = height x base. The large top opening makes it convenient for activities measuring mass by the displacement of water.
Customers Who Bought This Also Bought